siRNA Details
Gene Symbol: LARP4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015155.2 | NP_055970.1 | 18 | 5593 | ||
XM_017015987.1 | XP_016871476.1 | 17 | 7747 | |||
XM_017015988.1 | XP_016871477.1 | 17 | 7640 | |||
XM_017015989.1 | XP_016871478.1 | 17 | 7685 | |||
XM_017015992.1 | XP_016871481.1 | 13 | 4961 | |||
XM_017015993.1 | XP_016871482.1 | 16 | 5347 | |||
XM_017015994.1 | XP_016871483.1 | 17 | 5279 | |||
GenBank mRNA | BC003381.1 | - | 1 | 250 | ||
BC152443.1 | - | 17 | 5317 | 8862568 | ||
D86971.2 | - | 17 | 5317 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
