siRNA Details
Gene Symbol: FAM135B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015912.3 | NP_056996.2 | 4 | 407 | ||
XM_011517056.2 | XP_011515358.1 | 5 | 1213 | |||
XM_011517058.2 | XP_011515360.1 | 4 | 1068 | |||
XM_011517059.1 | XP_011515361.1 | 4 | 335 | |||
XM_011517060.2 | XP_011515362.1 | 5 | 1195 | |||
XM_011517061.2 | XP_011515363.1 | 5 | 552 | |||
XM_011517062.1 | XP_011515364.1 | 4 | 407 | |||
XM_011517063.2 | XP_011515365.1 | 4 | 1050 | |||
XM_011517064.2 | XP_011515366.1 | 5 | 1212 | |||
XM_011517065.1 | XP_011515367.1 | 4 | 471 | |||
XM_011517068.2 | XP_011515370.1 | 4 | 300 | |||
XM_011517070.2 | XP_011515372.1 | 5 | 1217 | |||
XM_017013470.1 | XP_016868959.1 | 4 | 1019 | |||
GenBank mRNA | AB196635.1 | - | 4 | 408 | ||
AK091433.1 | - | 4 | 407 | |||
BC037288.1 | - | 5 | 442 | 5266568 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
