siRNA Details
Gene Symbol: FAM135B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015912.3 | NP_056996.2 | 4 | 400 | ||
XM_011517056.2 | XP_011515358.1 | 5 | 1206 | |||
XM_011517058.2 | XP_011515360.1 | 4 | 1061 | |||
XM_011517059.1 | XP_011515361.1 | 4 | 328 | |||
XM_011517060.2 | XP_011515362.1 | 5 | 1188 | |||
XM_011517061.2 | XP_011515363.1 | 5 | 545 | |||
XM_011517062.1 | XP_011515364.1 | 4 | 400 | |||
XM_011517063.2 | XP_011515365.1 | 4 | 1043 | |||
XM_011517064.2 | XP_011515366.1 | 5 | 1205 | |||
XM_011517065.1 | XP_011515367.1 | 4 | 464 | |||
XM_011517068.2 | XP_011515370.1 | 4 | 293 | |||
XM_011517070.2 | XP_011515372.1 | 5 | 1210 | |||
XM_017013470.1 | XP_016868959.1 | 4 | 1012 | |||
XM_017013471.1 | XP_016868960.1 | 5 | 1206 | |||
GenBank mRNA | AB196635.1 | - | 4 | 401 | ||
AK091433.1 | - | 4 | 400 | |||
BC037288.1 | - | 5 | 435 | 5266568 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
