siRNA Details
Gene Symbol: FLJ38576 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046264.1 | 1 | 666 | |||
GenBank mRNA | AK095895.1 | - | 1 | 669 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
