siRNA Details
Gene Symbol: USF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009899.3 | NP_001009899.3 | 7 | 2145 | ||
XM_005247208.4 | XP_005247265.2 | 6 | 2073 | |||
XM_017005871.1 | XP_016861360.1 | 6 | 2040 | |||
XM_017005872.1 | XP_016861361.1 | 5 | 1957 | |||
GenBank mRNA | AB095938.1 | - | 1 | 58 | ||
CR749439.1 | - | 3 | 1688 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
