siRNA Details
Gene Symbol: TMED11P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033768.1 | 3 | 270 | |||
GenBank mRNA | AA887029.1 | - | 3 | 275 | ||
BG190838.1 | - | 1 | 103 | |||
BG215779.1 | - | 2 | 268 | |||
DY654917.1 | - | 1 | 104 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
