siRNA Details
Gene Symbol: HYDIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270974.2 | NP_001257903.1 | 37 | 5895 | ||
XM_006721206.2 | XP_006721269.1 | 37 | 5789 | |||
XM_011523146.2 | XP_011521448.1 | 38 | 5924 | |||
XM_011523147.1 | XP_011521449.1 | 38 | 5891 | |||
XM_011523148.1 | XP_011521450.1 | 38 | 5964 | |||
XM_011523151.1 | XP_011521453.1 | 37 | 5821 | |||
XM_017023346.1 | XP_016878835.1 | 37 | 6067 | |||
XM_017023347.1 | XP_016878836.1 | 25 | 4306 | |||
XM_017023348.1 | XP_016878837.1 | 25 | 4046 |
Gene Symbol: HYDIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103556.1 | 30 | 4852 | |||
GenBank mRNA | AB058767.1 | - | 4 | 584 | ||
AK027571.1 | - | 6 | 838 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
