siRNA Details
Gene Symbol: HYDIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270974.2 | NP_001257903.1 | 35 | 5454 | ||
XM_006721206.2 | XP_006721269.1 | 35 | 5348 | |||
XM_011523146.2 | XP_011521448.1 | 36 | 5483 | |||
XM_011523147.1 | XP_011521449.1 | 36 | 5450 | |||
XM_011523148.1 | XP_011521450.1 | 36 | 5523 | |||
XM_011523151.1 | XP_011521453.1 | 35 | 5380 | |||
XM_017023346.1 | XP_016878835.1 | 35 | 5626 | |||
XM_017023347.1 | XP_016878836.1 | 23 | 3865 | |||
XM_017023348.1 | XP_016878837.1 | 23 | 3605 |
Gene Symbol: HYDIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103556.1 | 28 | 4411 | |||
GenBank mRNA | AB058767.1 | - | 2 | 143 | ||
AK027571.1 | - | 4 | 397 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
