siRNA Details
Gene Symbol: LINC00663 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026956.1 | 3 | 464 | |||
GenBank mRNA | AK022418.1 | - | 1 | 98 | ||
AK289657.1 | - | 3 | 444 | |||
BC034236.1 | - | 4 | 596 | |||
DB196080.1 | - | 3 | 464 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
