siRNA Details
Gene Symbol: PSMA3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029434.1 | 1 | 102 | |||
NR_029435.1 | 1 | 102 | ||||
GenBank mRNA | AK055868.1 | - | 1 | 85 | ||
BC034618.1 | - | 1 | 94 | |||
BC104151.1 | - | 1 | 97 | |||
BC104152.1 | - | 1 | 97 | |||
BC112973.1 | - | 1 | 105 | |||
BC112974.1 | - | 1 | 102 | |||
BU533562.1 | - | 1 | 105 | |||
BX460997.2 | - | 1 | 87 | |||
BX482479.1 | - | 1 | 80 | |||
CR627086.1 | - | 1 | 95 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
