siRNA Details
Gene Symbol: CCDC184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013635.3 | NP_001013657.3 | 1 | 2147 | ||
GenBank mRNA | AK056003.1 | - | 1 | 2114 | ||
AK056783.1 | - | 1 | 1621 | |||
BC036801.1 | - | 1 | 2098 | 5728402 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
