siRNA Details
Gene Symbol: ITGAD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318185.1 | NP_001305114.1 | 9 | 1044 | ||
NM_005353.2 | NP_005344.2 | 9 | 1044 | |||
XM_011545835.2 | XP_011544137.1 | 9 | 1121 | |||
XM_011545836.2 | XP_011544138.1 | 9 | 1120 | |||
XM_011545837.2 | XP_011544139.1 | 9 | 1123 | |||
XM_011545838.2 | XP_011544140.1 | 9 | 1120 | |||
XM_011545839.2 | XP_011544141.1 | 9 | 1120 | |||
XM_011545841.2 | XP_011544143.1 | 9 | 1119 | |||
XM_011545842.2 | XP_011544144.1 | 9 | 1123 | |||
XM_011545843.2 | XP_011544145.1 | 9 | 1127 | |||
XM_011545844.2 | XP_011544146.1 | 9 | 1122 | |||
XM_011545845.2 | XP_011544147.1 | 9 | 1132 | |||
XM_011545846.2 | XP_011544148.1 | 9 | 1130 | |||
XM_011545847.2 | XP_011544149.1 | 9 | 1120 | |||
XM_011545848.2 | XP_011544150.1 | 9 | 1131 | |||
XM_017023215.1 | XP_016878704.1 | 9 | 1123 | |||
GenBank mRNA | AB208945.1 | - | 9 | 1044 | ||
AK097160.1 | - | 7 | 984 | |||
AK301028.1 | - | 8 | 1118 | |||
U37028.1 | - | 9 | 995 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
