siRNA Details
Gene Symbol: KIF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198525.2 | NP_940927.2 | 3 | 511 | ||
XM_011521531.2 | XP_011519833.1 | 3 | 621 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
