siRNA Details
Gene Symbol: GRAMD1C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172105.1 | NP_001165576.1 | 6 | 980 | ||
NM_017577.4 | NP_060047.3 | 11 | 1253 | |||
XM_005247546.2 | XP_005247603.1 | 10 | 1169 | |||
XM_005247547.1 | XP_005247604.1 | 11 | 1215 | |||
XM_011512930.1 | XP_011511232.1 | 11 | 1248 | |||
XM_011512931.1 | XP_011511233.1 | 6 | 1037 | |||
XM_017006646.1 | XP_016862135.1 | 10 | 1167 | |||
XM_017006647.1 | XP_016862136.1 | 12 | 1331 | |||
XM_017006648.1 | XP_016862137.1 | 7 | 1060 | |||
XM_017006649.1 | XP_016862138.1 | 5 | 811 | |||
GenBank mRNA | AK093181.1 | - | 6 | 772 | ||
AK097783.1 | - | 12 | 1337 | |||
AK292846.1 | - | 11 | 1189 | |||
AK292964.1 | - | 6 | 895 | |||
AK293884.1 | - | 9 | 965 | |||
AK304142.1 | - | 6 | 980 | |||
AL133661.1 | - | 2 | 1469 | |||
AY358725.1 | - | 6 | 933 | |||
BC028972.2 | - | 3 | 257 | |||
BC035040.1 | - | 11 | 1259 | 4829130 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
