siRNA Details
Gene Symbol: SLC4A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039960.2 | NP_001035049.1 | 25 | 8986 | ||
NM_001258401.2 | NP_001245330.1 | 25 | 8932 | |||
XM_011539010.1 | XP_011537312.1 | 26 | 9041 | |||
XM_011539011.1 | XP_011537313.1 | 26 | 8986 | |||
XM_011539014.2 | XP_011537316.1 | 16 | 7639 | |||
XM_017020240.1 | XP_016875729.1 | 26 | 9068 | |||
XM_017020242.1 | XP_016875731.1 | 26 | 9055 | |||
GenBank mRNA | AK124576.1 | - | 1 | 2184 | ||
AL137310.1 | - | 1 | 940 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
