siRNA Details
Gene Symbol: SLC4A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039960.2 | NP_001035049.1 | 25 | 10812 | ||
NM_001258401.2 | NP_001245330.1 | 25 | 10758 | |||
XM_011539010.1 | XP_011537312.1 | 26 | 10867 | |||
XM_011539011.1 | XP_011537313.1 | 26 | 10812 | |||
XM_011539014.2 | XP_011537316.1 | 16 | 9465 | |||
XM_017020240.1 | XP_016875729.1 | 26 | 10894 | |||
XM_017020242.1 | XP_016875731.1 | 26 | 10881 | |||
GenBank mRNA | AK124576.1 | - | 1 | 4010 | ||
AL137310.1 | - | 1 | 2766 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
