siRNA Details
Gene Symbol: C9orf106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012715.3 | NP_001012733.2 | 2 | 778 | ||
GenBank mRNA | AK092588.1 | - | 2 | 779 | ||
BC132699.1 | - | 2 | 775 | 40146721 | ||
BC132701.1 | - | 2 | 775 | 40146723 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
