siRNA Details
Gene Symbol: RBM44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080504.2 | NP_001073973.2 | 3 | 1059 | ||
XM_005246075.1 | XP_005246132.1 | 3 | 1415 | |||
XM_011511152.2 | XP_011509454.1 | 3 | 1415 | |||
XM_011511153.2 | XP_011509455.1 | 3 | 1415 | |||
XM_011511154.2 | XP_011509456.1 | 3 | 1415 | |||
XM_017004054.1 | XP_016859543.1 | 3 | 1415 | |||
XM_017004055.1 | XP_016859544.1 | 3 | 1785 | |||
XM_017004056.1 | XP_016859545.1 | 3 | 1355 | |||
XM_017004057.1 | XP_016859546.1 | 3 | 1415 | |||
GenBank mRNA | AK097730.1 | - | 3 | 1415 | ||
BX648107.1 | - | 3 | 994 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
