siRNA Details
Gene Symbol: LINC00282 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027047.1 | 5 | 3065 | |||
NR_027048.1 | 3 | 2922 | ||||
GenBank mRNA | AK094626.1 | - | 1 | 1611 | ||
AK124707.1 | - | 3 | 2922 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
