siRNA Details
Gene Symbol: MBLAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203397.2 | NP_981942.1 | 2 | 1391 | ||
XM_005250250.3 | XP_005250307.1 | 2 | 1434 | |||
GenBank mRNA | BC031288.1 | - | 2 | 1388 | 5299411 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
