siRNA Details
Gene Symbol: KIAA0895 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100425.1 | NP_001093895.1 | 3 | 296 | ||
NM_001199706.1 | NP_001186635.1 | 2 | 385 | |||
NM_001199707.1 | NP_001186636.1 | 2 | 537 | |||
NM_001199708.1 | NP_001186637.1 | 2 | 226 | |||
NM_015314.2 | NP_056129.2 | 2 | 393 | |||
XM_005249689.3 | XP_005249746.1 | 2 | 602 | |||
GenBank mRNA | AB020702.1 | - | 2 | 307 | ||
AK293912.1 | - | 2 | 158 | |||
AK294686.1 | - | 2 | 353 | |||
AK302559.1 | - | 2 | 353 | |||
BC028678.2 | - | 3 | 258 | 4836984 | ||
BC056191.1 | - | 2 | 277 | 5298453 | ||
BC110422.1 | - | 2 | 537 | 5199115 | ||
BC136362.1 | - | 2 | 287 | 9020349 | ||
BC144032.1 | - | 2 | 290 | 9052552 | ||
DB462178.1 | - | 3 | 298 | |||
DC403070.1 | - | 2 | 226 | |||
DC403204.1 | - | 2 | 385 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
