siRNA Details
Gene Symbol: KIAA0895 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100425.1 | NP_001093895.1 | 4, 5 | 1059 | ||
NM_001199706.1 | NP_001186635.1 | 4, 5 | 1292 | |||
NM_001199707.1 | NP_001186636.1 | 3, 4 | 1300 | |||
NM_001199708.1 | NP_001186637.1 | 4, 5 | 1133 | |||
NM_001300956.1 | NP_001287885.1 | 3, 4 | 444 | |||
NM_015314.2 | NP_056129.2 | 3, 4 | 1156 | |||
XM_005249689.3 | XP_005249746.1 | 4, 5 | 1509 | |||
XM_005249690.4 | XP_005249747.1 | 3, 4 | 606 | |||
GenBank mRNA | AB020702.1 | - | 3, 4 | 1070 | ||
AK293912.1 | - | 4, 5 | 1065 | |||
AK302559.1 | - | 4, 5 | 1260 | |||
AK302632.1 | - | 2, 3 | 236 | |||
BC028678.2 | - | 4, 5 | 1018 | 4836984 | ||
BC056191.1 | - | 3, 4 | 1040 | 5298453 | ||
BC136362.1 | - | 3, 4 | 1050 | 9020349 | ||
BC144032.1 | - | 4, 5 | 1197 | 9052552 | ||
BU507941.1 | - | 3, 4 | 346 | |||
DB060820.1 | - | 2, 3 | 467 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
