siRNA Details
Gene Symbol: DLGAP1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024101.1 | 3 | 532 | |||
GenBank mRNA | AK093095.1 | - | 3 | 532 | ||
BC137437.1 | - | 2 | 415 | |||
BX647607.1 | - | 1 | 2475 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
