siRNA Details
Gene Symbol: FLJ37453 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024279.1 | 2 | 1110 | |||
GenBank mRNA | AK094772.1 | - | 1 | 24 | ||
AK124018.1 | - | 2 | 1111 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
