siRNA Details
Gene Symbol: DPY19L2P4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003551.1 | 3 | 636 | |||
GenBank mRNA | BC031277.1 | - | 3 | 593 | ||
BC047471.1 | - | 3 | 584 | |||
BX537815.1 | - | 3 | 636 | |||
DQ287934.1 | - | 3 | 636 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
