siRNA Details
Gene Symbol: SOBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018013.3 | NP_060483.3 | 3 | 774 | ||
XM_005267041.4 | XP_005267098.1 | 3 | 782 | |||
XM_005267042.4 | XP_005267099.1 | 3 | 783 | |||
XM_011535920.2 | XP_011534222.1 | 3 | 782 | |||
XM_011535921.2 | XP_011534223.1 | 3 | 306 | |||
XM_017010991.1 | XP_016866480.1 | 2 | 1297 | |||
GenBank mRNA | AK090879.1 | - | 3 | 378 | ||
BC091526.1 | - | 3 | 283 | |||
BX442875.2 | - | 3 | 777 | |||
DQ507800.1 | - | 3 | 270 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
