siRNA Details
Gene Symbol: TRAPPC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042461.2 | NP_001035926.1 | 2 | 567 | ||
NM_001042462.1 | NP_001035927.1 | 2 | 419 | |||
NM_174894.2 | NP_777554.1 | 2 | 590 | |||
GenBank mRNA | AK292001.1 | - | 2 | 397 | ||
BC042161.1 | - | 2 | 536 | 4869377 | ||
BG528980.1 | - | 2 | 420 | |||
BI196928.1 | - | 2 | 511 | |||
BM841051.1 | - | 2 | 407 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
