siRNA Details
Gene Symbol: C5orf17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131245.1 | 3 | 642 | |||
GenBank mRNA | AK092155.1 | - | 3 | 642 | ||
BQ950913.1 | - | 3 | 602 | |||
DQ323997.1 | - | 3 | 417 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
