siRNA Details
Gene Symbol: TTC39C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243425.1 | NP_001230354.1 | 2 | 774 | ||
XM_005258198.4 | XP_005258255.1 | 2 | 257 | |||
XM_017025550.1 | XP_016881039.1 | 2 | 351 | |||
GenBank mRNA | AK074875.1 | - | 2 | 396 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
