siRNA Details
Gene Symbol: C1orf194 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006710342.3 | XP_006710405.1 | 1 | 535 | ||
XM_006710343.3 | XP_006710406.1 | 1 | 535 | |||
XM_011540647.2 | XP_011538949.1 | 1 | 536 | |||
XM_011540648.1 | XP_011538950.1 | 1 | 541 | |||
GenBank mRNA | BC127905.1 | - | 1 | 113 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
