siRNA Details
Gene Symbol: RPL17-C18orf32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199355.1 | NP_001186284.1 | 7 | 869 | ||
NM_001199356.1 | NP_001186285.1 | 7 | 936 | |||
GenBank mRNA | BX393840.2 | - | 7 | 871 |
Gene Symbol: C18orf32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035005.3 | NP_001030177.1 | 3 | 581 | ||
NM_001199346.1 | NP_001186275.1 | 3 | 417 | |||
GenBank mRNA | AB097012.1 | - | 3 | 501 | ||
AK027111.1 | - | 3 | 377 | |||
AK291882.1 | - | 3 | 388 | |||
BC022357.1 | - | 3 | 386 | 4297017 | ||
BC093004.1 | - | 3 | 538 | 5273608 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
