siRNA Details
Gene Symbol: C18orf32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035005.3 | NP_001030177.1 | 2 | 332 | ||
NM_001199346.1 | NP_001186275.1 | 2 | 168 | |||
GenBank mRNA | AB097012.1 | - | 2 | 252 | ||
AK027111.1 | - | 2 | 128 | |||
AK291882.1 | - | 2 | 139 | |||
BC022357.1 | - | 2 | 137 | 4297017 | ||
BC093004.1 | - | 2 | 289 | 5273608 |
Gene Symbol: RPL17-C18orf32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199355.1 | NP_001186284.1 | 6 | 620 | ||
NM_001199356.1 | NP_001186285.1 | 6 | 687 | |||
GenBank mRNA | BI461805.1 | - | 6 | 607 | ||
BX393840.2 | - | 6 | 622 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
