siRNA Details
Gene Symbol: LMO7DN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257995.1 | NP_001244924.1 | 1 | 112 | ||
XM_011535208.2 | XP_011533510.1 | 2 | 1939 | |||
GenBank mRNA | AK092698.1 | - | 1 | 113 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
