siRNA Details
Gene Symbol: LOC100049716 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122124.1 | 5 | 906 | |||
GenBank mRNA | AK056115.1 | - | 5 | 906 | ||
BC033133.1 | - | 2 | 1225 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
