siRNA Details
Gene Symbol: SYCN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080468.2 | NP_001073937.1 | 1 | 130 | ||
GenBank mRNA | AK130951.1 | - | 1 | 141 | ||
BC121075.2 | - | 1 | 134 | 40119665 | ||
BC121076.2 | - | 1 | 134 | 40119666 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
