siRNA Details
Gene Symbol: EIF4E1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099408.1 | NP_001092878.1 | 6, 7 | 937 | ||
XM_011534499.2 | XP_011532801.1 | 6, 7 | 1018 | |||
GenBank mRNA | AK094270.1 | - | 5, 6 | 1305 | ||
DR008159.1 | - | 3, 4 | 168 | |||
DR731203.1 | - | 3, 4 | 174 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
