siRNA Details
Gene Symbol: EIF4E1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099408.1 | NP_001092878.1 | 8 | 1087 | ||
XM_011534499.2 | XP_011532801.1 | 8 | 1168 | |||
GenBank mRNA | AK094270.1 | - | 6 | 1588 | ||
BM709255.1 | - | 3 | 175 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
