siRNA Details
Gene Symbol: ACER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010887.2 | NP_001010887.2 | 6 | 903 | ||
XM_005251448.3 | XP_005251505.1 | 7 | 954 | |||
XM_011517859.2 | XP_011516161.1 | 6 | 796 | |||
XM_017014694.1 | XP_016870183.1 | 6 | 659 | |||
GenBank mRNA | AF370405.1 | - | 9 | 1157 | ||
AK123581.1 | - | 6 | 903 | |||
AY312516.1 | - | 6 | 745 | |||
BC092487.1 | - | 7 | 954 | 30528463 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
