siRNA Details
Gene Symbol: ACER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010887.2 | NP_001010887.2 | 1 | 255 | ||
XM_005251448.3 | XP_005251505.1 | 1 | 170 | |||
XM_011517858.1 | XP_011516160.1 | 1 | 255 | |||
GenBank mRNA | AK123581.1 | - | 1 | 255 | ||
AY312516.1 | - | 1 | 97 | |||
BC092487.1 | - | 1 | 170 | 30528463 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
