siRNA Details
Gene Symbol: TBC1D22A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_122047.1 | 1 | 557 | |||
GenBank mRNA | AK057318.1 | - | 1 | 535 | ||
BC041833.1 | - | 1 | 557 | |||
CR456602.1 | - | 1 | 423 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
