siRNA Details
Gene Symbol: TFAP2A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033910.1 | 3 | 479 | |||
GenBank mRNA | AK092393.1 | - | 2 | 1043 | ||
BC012200.1 | - | 2 | 532 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
