siRNA Details
Gene Symbol: PSAPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085382.1 | NP_001078851.1 | 1 | 2203 | ||
GenBank mRNA | AK097698.1 | - | 2 | 1431 | ||
BC034552.1 | - | 1 | 2118 | |||
BC068579.1 | - | 1 | 2178 | |||
DQ991252.1 | - | 1 | 2203 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
