siRNA Details
Gene Symbol: ZBTB34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099270.1 | NP_001092740.1 | 2 | 1211 | ||
XM_005251989.4 | XP_005252046.1 | 3 | 1324 | |||
XM_011518699.2 | XP_011517001.1 | 2 | 1227 | |||
GenBank mRNA | AB082524.1 | - | 2 | 1211 | ||
DQ227306.1 | - | 1 | 1126 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
