siRNA Details
Gene Symbol: CMAHP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002174.2 | 2 | 226 | |||
NR_027626.1 | 3 | 545 | ||||
GenBank mRNA | AF074480.1 | - | 3 | 246 | ||
AK303297.1 | - | 3 | 502 | |||
BC032500.1 | - | 3 | 438 | |||
DA976765.1 | - | 3 | 546 | |||
FJ794466.1 | - | 2 | 204 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
