siRNA Details
Gene Symbol: CMAHP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002174.2 | 6, 7 | 872 | |||
GenBank mRNA | AF074480.1 | - | 7, 8 | 892 | ||
CR749466.1 | - | 5, 6 | 675 | |||
FJ794466.1 | - | 6, 7 | 850 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
