siRNA Details
Gene Symbol: CMAHP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002174.2 | 10 | 1433 | |||
GenBank mRNA | AF074480.1 | - | 11 | 1453 | ||
AK000716.1 | - | 4 | 2507 | |||
CR749466.1 | - | 9 | 1236 | |||
FJ794466.1 | - | 10 | 1411 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
