siRNA Details
Gene Symbol: SHISA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080505.1 | NP_001073974.1 | 2 | 957 | ||
GenBank mRNA | BC012029.1 | - | 1 | 457 | ||
BC127690.1 | - | 2 | 798 | 40127704 | ||
BC127691.1 | - | 2 | 798 | 40127705 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
