siRNA Details
Gene Symbol: FAM86JP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024250.1 | 5 | 809 | |||
NR_024251.1 | 5 | 907 | ||||
GenBank mRNA | BC032772.1 | - | 5 | 795 | ||
BC041080.1 | - | 5 | 878 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
