siRNA Details
Gene Symbol: FAM86DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | DA753838.1 | - | 3, 4 | 345 |
Gene Symbol: FAM86C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099653.1 | NP_001093123.1 | 3, 4 | 267 | ||
GenBank mRNA | AK311428.1 | - | 3, 4 | 244 | ||
AK311518.1 | - | 3, 4 | 267 | |||
BQ921333.1 | - | 3, 4 | 237 |
Gene Symbol: FAM86JP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024250.1 | 3, 4 | 264 | |||
NR_024251.1 | 3, 4 | 362 | ||||
GenBank mRNA | BC032772.1 | - | 3, 4 | 250 | ||
BC041080.1 | - | 3, 4 | 333 | |||
BM781991.1 | - | 3, 4 | 264 | |||
DA599453.1 | - | 3, 4 | 533 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
