siRNA Details
Gene Symbol: MUC3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005960.1 | NP_005951.1 | 10 | 10044 | ||
XM_011516246.2 | XP_011514548.1 | 11 | 10373 | |||
XM_011516247.2 | XP_011514549.1 | 11 | 10323 | |||
XM_011516249.2 | XP_011514551.1 | 11 | 10174 | |||
XM_011516250.2 | XP_011514552.1 | 11 | 9662 | |||
XM_017012231.1 | XP_016867720.1 | 11 | 9870 | |||
GenBank mRNA | AB038784.1 | - | 9 | 2501 | ||
AF007194.1 | - | 10 | 3403 | |||
AF143371.1 | - | 8 | 968 | |||
AF143372.1 | - | 7 | 794 | |||
AF143373.1 | - | 7 | 852 |
Gene Symbol: MUC3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB038783.1 | - | 9 | 2504 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
